missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Square and Rectangular Cover Glasses
Provides quality, reliability and performance for your laboratory microscopy needs
Brand: Corning™ 2975-244
This item is not returnable.
View return policy
Description
Provides quality, reliability and performance for your laboratory microscopy needs
- Made from soda-lime glass and borosilicate glass with high clarity
- Available in square and rectangular shapes, and in three standard sizes and thicknesses
Order Info
Packaged in plastic boxes for protection during storage.
Specifications
Coverslip | |
Borosilicate Glass | |
24 mm | |
Non-sterile | |
100/Pk. |
Rectangle | |
40 mm | |
#1 (0.13 to 0.16 mm) | |
Microscopes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction