Learn More
Nabertherm™ Square Saggar
Square saggar for furnaces LHTC and LH.
Marca: Nabertherm™ 699000279
Code : NEW
Detalles adicionales : Weight : 0.50000kg
Descripción
- Tmax: 1600°C
The load is placed in ceramic saggars for optimal utilization of the furnace space. Up to three saggars can be stacked on top of each other in the furnace. Each saggar has cut-outs for better ventilation. The top saggar should be closed with a lid made of ceramics also.
Especificaciones
1600°C | |
Square Saggar |