missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SR 3576
Highly potent and selective JNK3 inhibitor
£133.00 - £502.00
Specifications
Purity | >97% |
---|---|
Product Type | SR 3576 |
Molecular Weight (g/mol) | 501.53 |
Inhibitors | SR 3576 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18761779
|
Tocris Bioscience™
5044/10 |
10 mg |
£133.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18747048
|
Tocris Bioscience™
5044/50 |
50 mg |
£502.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Highly potent and selective JNK3 inhibitor (IC50 = 7 nM); exhibits > 2800-fold selectivity over p38.Specifications
>97% | |
SR 3576 | |
501.53 | |
SR 3576 |
For Research Use Only