missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRP19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16033-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is a rabbit polyclonal antibody raised against full length SRP19 of human origin.
SRP19 belongs to the SRP19 family. It is signal-recognition-particle assembly and binds directly to 7S RNA and mediates binding of the 54 kDa subunit of the SRP.Specifications
SRP19 | |
Polyclonal | |
Unconjugated | |
SRP19 | |
SRP19 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
291685, 66384, 6728 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P09132, Q9D7A6 | |
Srp19 | |
SRP19 Fusion Protein Ag8903 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |