missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSC 20x Buffer, MP Biomedicals™
Click to view available options
Quantity:
500 mL
Description
- SSC Buffer used in electrophoresis and in sedimentation studies of DNA-histone aggregates.
- SSC is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.
Specifications
Physical Form | Solution |
Concentration | 20× Concentrated |
Color | Clear Colorless |
Quantity | 500 mL |
Grade | Molecular Biology |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'