missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals SSC buffer (20X)

Suitable for preparing nucleic acids for hybridization, hybridization buffer for northern and southern blots, washing buffer for northern and southern blots. Saline-Sodium Citrate Buffer (SSC) is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.

Brand:  Thermo Scientific Chemicals J60839.K2

40.10 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15413549

  • £40.30 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

SSC buffer is suitable for preparing nucleic acids for hybridization, hybridization buffer for northern and southern blots, washing buffer for northern and southern blots. Saline-Sodium Citrate Buffer (SSC) is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ambient temperatures
Liquid
1 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN14L550 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog NumberJ60839.K2,J60839.K3,J60839.K7
Lot NumberN14L549 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog NumberJ60839.K2,J60839.K3,J60839.K7
Lot NumberX28K521 Certificate TypeCertificate of Analysis Date14/11/2024 Catalog NumberJ60839.K2,J60839.K3,J60839.K7
Lot NumberW26K564 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog NumberJ60839.K2,J60839.K3,J60839.K7
Lot NumberR24K511 Certificate TypeCertificate of Analysis Date01/07/2024 Catalog NumberJ60839.K2,J60839.K3,J60839.K7
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals SSC buffer (20X) > 1L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only