missing translation for 'onlineSavingsMsg'
Learn More

SST Clot Activator and Polymer Gel Hemogard™ Closure Blood Tube

Use the SST Clot Activator and Polymer Gel Hemogard™ Closure Blood Tube, which contains Silica, to activate clotting of the specimen and a gel that forms a barrier between the clot and the serum after centrifugation.

Brand:  BUNZL Healthcare VS367954

Product Code. 12336939

  • £182.00 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Volume - 5ml
  • Additives - Silica (Clot Activator)/Gel
  • Cap Color - Gold
  • Size (mm) - 13x100
  • Quantity - 100

Pack of 100
Specifications

Specifications

Blood Collection Tube
Hemogard
Gold
13 mm
SST Clot Activator, Polymer Gel
5 mL
100 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.