missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Assistent™ ThermoClean™ DC Stabilizing Solution

Product Code. p-8007140
Click to view available options
Color:
Clear
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 10449591

Brand: Karl Hecht™ 43170003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Prevents algae/bacterial growth and stops odor formation in water baths. Karl Hecht™ Assistent™ ThermoClean™ DC Stabilizing Solution is suitable for all kinds of thermostats and circulation pumps made of metal, glass or any plastics.

  • Solvent-free
  • Only minor pH-change to the water
  • Prevents foam formation, even with strong circulation
  • No obstructions in tubings and temperature control systems
  • Can also be used for closed temperature control systems
  • Free of aggressive phosphates and oxidizing or reducing substances
  • Available in blue and clear variants 

Specifications

Color Clear
Type Stabilizing Solution
Quantity 100 mL
compliance-icons
Product Identifier
  • Stabilising Solution for Water Baths ThermoClean DC
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : benzyl-C12-16-alkyldimethyl, chlorides,Poly (N,N-Dimethyl-2-hidroxypropyl) ammonium Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.