Learn More
Hach™ Stablcal™ Turbidity Standards Calibration Kit, Sealed Vials
Developed for use in any turbidimeter. Hach™ Stablcal™ Turbidity Standards Calibration Kit, Sealed Vials are true Formazin dilutions in individual sealed vials of <0.1, 20, 200, 1000, 4000 NTU and 7500 NTU (2662105 only) levels.
£459.00
Specifications
Type | Stablcal Turbidity Standards Calibration Kit |
---|---|
Includes | Premixed, sealed vials |
For Use With (Application) | Calibration of turbidimeters |
Product Code | Brand | Turbidity | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Turbidity | For Use With (Equipment) | Price | Quantity & Availability | ||||
10070654
|
Hach™
2662105 |
<0.1, 20, 200, 1000, 4000 NTU | 2100N/N IS |
£459.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Developed for use in any turbidimeter. Hach™ Stablcal™ Turbidity Standards Calibration Kit, Sealed Vials are true Formazin dilutions in individual sealed vials of <0.1, 20, 200, 1000, 4000 NTU and 7500 NTU (2662105 only) levels.
- Precise, Pre-mixed Formazin Solutions: Provides accurate turbidimeter calibrations
- Based on Formazin Primary Standards: Contains the same light scattering polymer and performs equivalently
- Enhanced Stability Saves Time and Labor: Repeatable results without preparing standards in the lab
- Confidence in Results: Based on decades of experience in the drinking water industry
- Any Turbidity Meter: Can be used with 2100N/2100AN or 2100N/2100AN IS Turbidimeter
Specifications
Stablcal Turbidity Standards Calibration Kit | |
Premixed, sealed vials | |
Calibration of turbidimeters |
Safety and Handling
- StablCal sealed vials calibration kits
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P363-Wash contaminated clothing before reuse.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
- MIXTURE LIST-Contains : methenamine; hexamethylenetetramine, methenamine; hexamethylenetetramine , methenamine; hexamethylen
Your input is important to us. Please complete this form to provide feedback related to the content on this product.