Learn More
Cole-Parmer™ Stackable HDPE Bucket
Stackable buckets have a tapered body to permit one nesting inside the other for easy storage. They feature a bail-type handle and recessed fingergrip rim. Standard lid and lid with pour spout are available but not included.
Brand: Cole-Parmer™ 06274-25
This item is not returnable.
View return policy
Description
- Stackable - saves space
- Recessed grip handle
- Lids available separately
- High density polyethylene
Specifications
Not Autoclavable |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Cole-Parmer™ Stackable HDPE Bucket > Bucket; Capacity: 19 L; Material: HDPE
Spot an opportunity for improvement?Share a Content Correction