Learn More
Gilac™ Stackable Reinforced Crate with Handles
Large reinforced storage container in food-grade plastic (HDPE). Stackable bin with multi-purpose handles, ideal for handling.
Brand:  Gilac™ G154621
 View more versions of this product
Product Code. 30245683
  • £31.44 / Each
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Large reinforced storage container in food-grade plastic (HDPE)
  • Stackable bin with multi-purpose handles, ideal for handling
  • Ideal for negative or positive cold storage
  • This reinforced, professional-quality bin is designed for intensive use
Specifications
Specifications
Storage Container
White
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gilac™ Stackable Reinforced Crate with Handles > Gilac™ Reinforced Crate, Color, White