Learn More
BINDER™ Stacking Adapter
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB 65
KB/KB PRO 130, KBF/KBF PRO 130
Unit Size:
Each
Description
- Used to safely stack two identical devices
- For use with KB 65, KB 130, KB PRO 130, KBF 130, or KBF PRO 130 models
Specifications
Specifications
| For Use With (Equipment) | KB/KB PRO 130, KBF/KBF PRO 130 |
| Material | Plastic |
| Type | Stacking Adapter |