Learn More
Eppendorf™ Stacking stand set
For Use With: CellXpert™ C170/C170i Incubators
Brand: Eppendorf™ 6731080005
993.26 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 20.00000kg
Description
- WhatsNext 2019 Bundle: Stacking stand set for CellXpert™ C170/C170i incubator, consisting of one lower stacking stand with castors and one upper stacking stand
Specifications
CellXpert™ C170/C170i Incubators | |
Stacking Set | |
One lower stacking stand with castors, and one upper stacking stand |