missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Stained Salmonella O and H Suspensions

Rapidly detect and quantify specific antibodies to Salmonella in sera with this standardized stained suspension.

Brand:  Thermo Scientific™ R30855401

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11913272

  • £124.00 / 5mL
Estimated Shipment: 23-07-2024
to see stock.

Description

Description

Rapidly detect and quantify specific antibodies to Salmonella spp. in sera with Thermo Scientific™ Remel™ Stained Salmonella O and H Suspensions. Stained Salmonella O and H Suspensions are standardized, smooth suspensions of killed bacteria which have been stained to facilitate reading of agglutination tests.

Use Stained Salmonella O and H Suspensions for the qualitative detection of Salmonella antibodies in sera for epidemiological and diagnostic purposes, primarily in the investigation of enteric infections and pyrexia.

  • Identifies major groups of Salmonella serotypes
  • Provides accurate, reliable results
  • Simple, easy-to-use tests

Serological tests are based on the fact that antibodies in serum, produced in response to exposure to bacterial antigens, will agglutinate bacterial suspensions which carry homologous antigens. In the diagnosis of febrile disease such tests should be used in parallel with appropriate cultural techniques for the isolation and identification of the causative organism. The suspensions are suitable for use in rapid slide tests1 and standard Widal tests2, 3. They may also be used as control antigens for serological identification of bacterial isolates.

 Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.

Specifications

Specifications

2°C to 8°C
S. paratyphi B-O
5 mL
Stained Salmonella O and H Suspensions
Stained Salmonella O and H Suspensions, instruction for use
Salmonella
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers