Learn More
Brady™ StainerBondz™ Microscope Slide Matte Polyester Labels
Easily create chemical-resistant microscope slide labels with the StainerBondz™ polyester label material (B-481). Print almost anywhere with a Brady™ handheld label printer.
Brand:  Brady™ 170885
 View more versions of this product
Product Code. 17871719
  • £49.25 / Pack of 320
Explore more special offers
Description
Description
  • Available in presized shapes for commonly used slides and tissue cassettes in which superior chemical resistance is required
  • Withstands the rigors of typical deparaffinization including ethanol and xylene, as well as hematoxylin and eosin slide staining processes
  • Made with a permanent acrylic adhesive for tight bonding onto smooth surfaces in extreme conditions
Specifications
Specifications
Autoclavable
M5-195-481
None
Matte
Polyester
Thermal Transfer
Non-sterile
BMP51, M511
-112°F to +266°F
0.375 x 1 in.
1 in.
0.5 in.
RoHS Compliant to 2011/65/EU (Pass 2015/863)
Permanent Acrylic
Black on White
Die-cut
Top
Labels
Healthcare and Hospital Labeling, General Identification, Laboratory Labeling
320 Labels/Cartridge
0.0033 in.
0.375 in.
2.54 cm