missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sampling Systems™ Stainless Dipper 600mm Long
Designed to be easy to use and suitable for a range of liquids. Sampling Systems™ Stainless Dipper 600mm Long is ideal for obtaining a surface sample from a wide range of environments.
Brand: Sampling Systems™ 5005A-100
This item is not returnable.
View return policy
Description
- Manufactured in 316L Stainless Steel
- Fully welded construction
- Satin finish
- Easy to use
- For liquid sampling
Specifications
Dipper | |
100 mL | |
600 mm |
Stainless Steel (316L) | |
39 mm |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sampling Systems™ Stainless Dipper 600mm Long > Capacity, Metric: 100mL
Spot an opportunity for improvement?Share a Content Correction