missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sampling Systems™ Stainless Dipper 600mm Long
Designed to be easy to use and suitable for a range of liquids. Sampling Systems™ Stainless Dipper 600mm Long is ideal for obtaining a surface sample from a wide range of environments.
Brand: Sampling Systems™ 5005A-250
This item is not returnable.
View return policy
Description
- Manufactured in 316L Stainless Steel
- Fully welded construction
- Satin finish
- Easy to use
- For liquid sampling
Specifications
Dipper | |
250 mL | |
600 mm |
Stainless Steel (316L) | |
65 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction