Learn More
Metro™ Stainless Lab Worktable, Stainless Island Top and 3-Sided Frame
Keep lab materials clean and safe. Metro Stainless Lab Worktable features Type 304 stainless steel construction throughout and 3-sided Frame providing corrosion-proof performance and stability you can count on.
Brand:  Metro™ LTSM36UIS
 View more versions of this product
Product Code. 15957131
  • £1860.00 / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Specifications
Mobile Table
Ideal for lab
89 cm
35.75 in.
600 lb.
30 in.
Stainless Steel
35.75 x 30 x 35 in.
35 in.
3 Sided frame, adaptor insert
90.8 cm
273 kg
76.2 cm