missing translation for 'onlineSavingsMsg'
Learn More

Fritsch™ Stainless Steel 1.4404/316L Sieve Ring

Brand:  Fritsch™ 44.2100.10

Code : 04

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 13121674

  • POA / Each
Estimated Shipment: 18-09-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

Specifications

Specifications

PULVERISETTE 14 Variable Speed Rotor Mill
Stainless Steel (316L)
1 mm
Round holes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers