missing translation for 'onlineSavingsMsg'
Learn More

Fritsch™ Stainless Steel 1.4404/316L Sieve Ring

Brand:  Fritsch™ 44.1050.10

Code : 04

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 13137893

  • POA / Each
Estimated Shipment: 18-09-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

Specifications

Specifications

PULVERISETTE 14 Variable Speed Rotor Mill
Stainless Steel (316L)
0.5 mm
Trapezoid holes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers