missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ BinLiner™ Stainless Steel BinStand™

These two styles of bin liner stands are made from durable, stainless steel, open rod frames that allow ergonomic side removal of liners for simplified use and securely hold a range of bin liner bags. The perfect compliment to Micronova BinLiners™.

Brand:  Micronova™ BLSSH-1515

 View more versions of this product

Product Code. 17990050

  • £269.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The scissor style stainless steel frame bag holders designed to hold 40 inch x 48 inch BinLiners™, for use in clean and controlled environments.

The Micronova BinLiner™ Stainless Steel BinStands™ are ideal for cleanroom material disposal, garment collection and raw material or product storage. The unique, rectangular stand features polyketone molds that connect the sides and function as stand feet that easily slide to prevent floor scratching and rusting. The scissor stand easily folds open and closed for storage when not in use.

  • Two styles of stainless steel rod, open frame bin stands
  • Ergonomic side removal of liners for simplified use
  • Securely hold a range of bin liner bags
  • For material disposal, garment collection and raw material or product storage
  • The scissor stand easily folds open and closed for storage
  • Rectangular stand has plastic connectors that prevent floor rusting and scratching
  • Corner legs are removable if disassembly is required
  • Available in a variety of sizes to hold bags from 5 to 55 gallon sizes
    • BLSSH-1515 stand for use with 34 x 40-inch liners
    • BLSSH-1624 stand for use with 40 x 48-inch and 40 x 60-inch liners
    • BLSSH-40 stand best used with 40 gallon liners
  • Resistant to common cleaning solutions and disinfectants
  • Autoclavable
Specifications

Specifications

120 L
30 gal.
71 cm
1 Ea.
Autoclavable
For Use with 34 x 40-Inch Liners
15 in.
Stainless Steel
Stainless Steel Frame Bag Holder
38 x 38 x 71 cm (15 x 15 x 28 in.)
38 cm
38 cm
Stand for Disposal Bags Used in Controlled Environments
15 in.
28 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number047872-000 Certificate TypeCertificate of Compliance Date31/01/2025 Catalog NumberBLSSH-1515
Lot Number046049-000 Certificate TypeCertificate of Compliance Date01/11/2024 Catalog NumberBLSSH-1515
Lot Number046154-000 Certificate TypeCertificate of Compliance Date15/10/2024 Catalog NumberBLSSH-1515
Lot Number045109-000 Certificate TypeCertificate of Compliance Date16/08/2024 Catalog NumberBLSSH-1515
Lot Number044832-000 Certificate TypeCertificate of Compliance Date09/08/2024 Catalog NumberBLSSH-1515
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Micronova™ BinLiner™ Stainless Steel BinStand™ > Wire BinLiner Stand, Stainless Steel with plastic corners, 15 x 15 x 28 in., 120 L (30 gal.)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.