Learn More
Grant Instruments™ Stainless Steel Bath Cover
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Bath Cover for Grant Stirred Water Baths is available flat or gabled with removable hinge.
Brand:  Grant Instruments™ STL38
 View more versions of this product
Product Code. 12777779
  • £608.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Bath Cover for Grant Stirred Water Baths is available flat or gabled with removable hinge.
  • Stainless steel construction
  • Plastic handle
  • Available flat (STL 5), or gabled with removable hinge (STL 12, STL 38)
  • For use with Grant Stirred Water Baths and Heated Circulators
  • Prevents heat from escaping
  • Reduces sample contamination
Specifications
Specifications
ST38 tank
Stainless Steel