Learn More
Grant Instruments™ Stainless Steel Bath Cover
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Bath Cover for Grant Stirred Water Baths is available flat or gabled with removable hinge.
Brand: Grant Instruments™ STL5
Description
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Bath Cover for Grant Stirred Water Baths is available flat or gabled with removable hinge.
- Stainless steel construction
- Plastic handle
- Available flat (STL 5), or gabled with removable hinge (STL 12, STL 38)
- For use with Grant Stirred Water Baths and Heated Circulators
- Prevents heat from escaping
- Reduces sample contamination
Specifications
ST5 tank | |
Stainless Steel |