Learn More
Stainless Steel Blunt End Spring Forceps
Use these forceps for a variety of life science applications. S Murray™ Stainless Steel Blunt End Spring Forceps are fully guaranteed in normal use and subject to stringent quality inspection procedures.
Brand:  Samco E003/05
Product Code. 12750916
  • £23.70 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
Use these forceps for a variety of life science applications. S Murray™ Stainless Steel Blunt End Spring Forceps are fully guaranteed in normal use and subject to stringent quality inspection procedures.
  • Standard instruments are finished to a bright mirror polish
  • Stainless Steel
  • Length: 200mm
Specifications
Specifications
Spring Forceps
For dissecting
200 mm
Stainless Steel
Blunt