missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Stainless Steel Bosshead

Product Code. 13489618
Click to view available options
Color:
Silver
Unit Size:
Each

Product Code. 13489618

Brand: Bochem™ 5354

Only null left
Add to basket
Add to basket

Bochem™ bossheads are manufactured from 18/10 stainless steel, with 180° and 90° angle, and M8 'heart' safety screw.

  • 18/10 stainless steel
  • Safety screw type 'heart' M8
  • 180+90° angle

Specifications

Type Bosshead Clamp
Material Stainless Steel
Includes Safety Screw Type 'Heart' M8
For Use With (Equipment) Rods and Stand Bases
For Use With (Application) Holding and Supporting Laboratory Apparatus
Diameter (Metric) 16.5 mm
Color Silver
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.