Learn More
Memmert™ Stainless Steel Bottom Drip Tray for SN750, 15 mm
Non-perforated bottom drip trays constructed of stainless steel.
Brand: Memmert™ SN750B04362
Additional Details : Weight : 0.01200kg
Description
- Accessory
- May affect temperature distribution
- Constructed of stainless steel
- Rim height: 15 mm
Specifications
SN750 Sterilizer | |
Bottom Drip Tray | |
Rim: 15 mm | |
Stainless Steel |