Learn More
Sampling Systems™ Stainless Steel Bucket
Sampling Systems™ Stainless Steel Buckets are made from a heavy gauge 316L Stainless Steel for durability.
Brand:  Sampling Systems™ A586-12
 View more versions of this product
Product Code. 15819701
  • £210.00 / Each
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made from 316L Stainless Steel
  • Crevice free interior
  • Swing handle
  • Flat base