Learn More
Nickel Electro™ Stainless Steel Buckets without Graduations
Quick to clean and having an attractive shiny appearance, this Nickel Electro™ Stainless Steel Bucket without Graduation, provides good chemical resistance and can handle hot liquids.
Brand:  Nickel Electro™ BUCS-0100
 View more versions of this product
Product Code. 12802034
  • £109.00 / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • High quality stainless steel construction without graduations
  • Bright polished finish for easy cleaning
  • Good chemical resistance
  • Can be autoclaved
Specifications
Specifications
Bucket
Stainless Steel
Stainless steel
Bright Polished
10 L
Autoclavable
1 Ea.