Learn More
Sampling Systems™ Stainless Steel Buckets with Skirt Band and Graduations
Sampling Systems™ Stainless Steel Buckets with Skirt Band and Graduations are made from heavy gauge, durable stainless steel and are ideal for hygienic applications.
Brand:  Sampling Systems™ A264-10
Additional Details : Weight : 4.68000kg
 View more versions of this product
Product Code. 15859601
  • £92.30 / Each
Estimated Shipment: 18-07-2024
to see stock.
Add to basket
Description
Description
  • Crevice free interior
  • Graduated
  • Supplied with skirt band
  • Robust design
  • 304 Stainless Steel