Learn More
VACUUBRAND™ Stainless Steel Butterfly Passage Valve
Product Code. p-8007644 Shop All Vacuubrand Products
Click to view available options
For Use With:
KF VS 40/ DN
KF VS16 / DN
KF VS25 / DN
Unit Size:
Each
Product Code. 10438352
Brand: VACUUBRAND™ 20665004
Only null left
Add to basket
Add to basket
Compact, economic design, easy to grip, lever-operated with position indicator
The VS und VS C series valves are butterfly-type valves. A circular valve plate with a sealing ring on the circumference rotates around an axis at right angle to the valve axis. As a result, the valves leave virtually the entire cross section free, offering very good conductance. The valve bodies are made of one piece, machined from solid material, and require (in addition to the valve head seal) only one elastic shaft seal for rotational movement of the shaft. The valves provide low leakage and minimal degassing rate. The VS C series features a valve seal made of perfluorelastomer material (ffKM).
TRUSTED_SUSTAINABILITY
Specifications
For Use With KF VS16 / DN
Material Stainless Steel
Quantity 1 Each
Type Butterfly Passage Valve