Learn More
Thermo Scientific™ Stainless-Steel Cabinet Finishes
Brand:  Thermo Scientific™ 6116
Product Code. 15108344
  • £1330.72 / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Stainless-Steel Cabinet Finishes are for Thermo Scientific High-Performance Lab Refrigerators and Freezers.
  • Optional stainless steel interior finish
  • Adds 1in. or 25mm to unit height
  • Must be specified when ordering
  • Options available only on certain models
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless-Steel Interior Finish
51.1 cu. ft. cabinets