Learn More
Ohaus™ Stainless Steel Calibration Weight
Adjusted to meet or exceed OIML M1 tolerances
Brand:  Ohaus™ 11117146
27.08 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 12773285
  • £29.65 / Each
Estimated Shipment: 20-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Meets OIML M1 standards
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OIML M1 class
50 g