Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML E2
Adjusted to Meet or Exceed OIML E2 Tolerances
Brand: Ohaus™ 30518023
This item is not returnable.
View return policy
Description
- Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
No Certificate | |
2 kg | |
Protective Case |
Troemner Alloy 8 | |
3 mg |