missing translation for 'onlineSavingsMsg'
Learn More

3M™ Stainless Steel Cleaner & Polish Aerosol

Stainless Steel Cleaner & Polish Aerosol

Brand:  3M™ 7000116804

Additional Details : Weight : 7.20000kg Transport : UN number : 1950 Chem class : 2.1


Product Code. 17227630

  • £233.00 / 12 canisters
Estimated Shipment: 14-08-2024
to see stock.
Add to basket

Description

Description

  • Polishes and protects all stainless steel surfaces in one step
  • The spray clings to vertical surfaces and leaves a light, non-greasy film that resist fingerprints and marking
  • Does not contain harmful chemicals and safe to use in food preparation areas
  • Water based with a pleasant citrus fragrance
Specifications

Specifications

Cleaner
600 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3M Stainless Steel Cleaner

Signal Word
  • Danger

Hazard Category
  • Flammable and non-flammable aerosols category 2

Hazard Statement
  • H223-Flammable aerosol.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P211-Do not spray on an open flame or other ignition source.
  • P251-Do not pierce or burn, even after use.
  • P410+P412-Protect from sunlight Do no expose to temperatures exceeding 50°C/122°F.

Supplemental information
  • MIXTURE LIST-Contains : butane, isobutane, propane

SDS
Documents

Documents

Certificates
Special Offers

Special Offers