Learn More
Nickel Electro™ Stainless Steel Flask Clip
Product Code. p-8013893 Shop All Nickel-Electro Products
Click to view available options
Holds:
1 L Conical Flask
100 mL Conical Flask
2 L Conical Flask
2.8 L Fernbach flask
25 mL Conical Flask
250 mL Conical Flask
50 mL Conical Flask
500 mL Conical Flask
Unit Size:
Each
Product Code. 11479629
Brand: Nickel Electro™ FC1000
Only null left
Add to basket
Add to basket
For personalised clip arrangement, clips are supplied with all necessary fixings to enable them to be attached to the Perforated Tray. Nickel Electro™ Stainless Steel Flask Clip is suitable for flasks and can be autoclaved.
  • Highly flexible elastic design allowing glassware to easily slide into the clip and yet remain secure during mixing
  • No sharp edges to cut or score glassware
  • Can be autoclaved
  • Plastic base
  • Easy to assemble and remove Flask Clips from Perforated tray
Specifications
Holds 1 L Conical Flask
Product Type Flask Clip
Material Stainless Steel