missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Stainless Steel Comfort Rack for Upright Freezers

Place the cryoboxes in drawers with the Tenak™ Stainless Steel Comfort Rack for Upright Freezers .This comfort rack structure increases the flexibility and reduces the operating weight when compared to rack types.

Brand:  Tenak™ TE24232

 View more versions of this product

Product Code. 15160875

  • £152.00 / Each
Estimated Shipment: 02-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Six different box heights
  • Fit all freezer models
  • Stainless steel.
  • Cryoboxes sold separately
Specifications

Specifications

Yes
4 x 4
Stainless Steel
Upright Freezers
1 Pack
140 mm
558 mm
Comfort Rack
Boxes (4 x 4/16)
16
16
1 Pack
228 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tenak™ Stainless Steel Comfort Rack for Upright Freezers > Holds: 4 x 4/16 Boxes; Height: 228mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.