missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Stainless Steel Container
Bochem™ Stainless Steel Container is constructed of Stainless Steel and is equipped with handles on the side.
£511.00 - £1220.00
Product Code | Brand | Diameter (Metric) Inner | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
10342711
|
Bochem™
8340 |
270mm | 20 L | 375mm |
£511.00
Each |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||
10020811
|
Bochem™
8346 |
400mm | 75 L | 600mm |
£1025.00
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||
10761962
|
Bochem™
8341 |
300mm | 25 L | 375mm |
£601.00
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||
10172791
|
Bochem™
8347 |
450mm | 100 L | 670mm |
£1220.00
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||
Description
- Stainless Steel
- Includes handles
- Compatible lid sold separately
Specifications
Stainless Steel (18/10) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bochem™ Stainless Steel Container