Learn More
AMSTA Rostfritt Stainless Steel Container with Lid
High-quality stainless steel container, available in various sizes with or without clamps and tap.
Brand:  AMSTA Rostfritt 50608251+5051310X3
 View more versions of this product
Product Code. 11741944
  • £1180.00 / Each
Explore more special offers
Description
Description
  • Quality
  • Invisible weld seams
  • Pharmaceutical grade finish
  • Sustainable production
  • Made in Sweden
Specifications
Specifications
Can
Silver
Ra 0.2 to 0.3 μm
397 mm
4.8 kg
25 L
Stainless Steel
Lid and 3 Clamps
300 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products