Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class E2
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
£838.24 - £3335.00
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | ||||||
15565119
|
Fisherbrand™
FB67837 |
200 g |
£1020.00
1 unit |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
15565129
|
Fisherbrand™
FB67847 |
1 g to 5 kg |
£3335.00
1 unit |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
15585119
|
Fisherbrand™
FB67839 |
1 g to 1 kg |
£1615.00
1 unit |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
15505109
|
Fisherbrand™
FB67821 |
1 to 100 g |
£838.24
1 unit |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
Description
- Class E2 stainless steel calibration weight
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
E2 | |
Cylindrical |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class E2