Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class F2 with certificate
Use with weighing instruments of high accuracy class II. Fisherbrand™ Class F2 Stainless Steel Calibration Weights are also intended for use in important commercial transactions, such as those involving precious metals and stones.
£465.00 - £1865.00
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | ||||||
15535129
|
Fisherbrand™
FB67844 |
1 g to 1 kg |
£976.00
1 unit |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
15535119
|
Fisherbrand™
FB67834 |
1 to 100 g |
£465.00
1 unit |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
15515139
|
Fisherbrand™
FB67852 |
1 g to 5 kg |
£1865.00
1 unit |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
Description
- Class F2 stainless steel calibration weight
- Weights intended for use in the verification or calibration of class M1 and possibly class M2 weights
- Delivered with a calibration certificate guaranteeing traceability
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
F2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class F2 with certificate