Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class F2
Use with weighing instruments of high accuracy class II. Fisherbrand™ Class F2 Stainless Steel Calibration Weights are also intended for use in important commercial transactions, such as those involving precious metals and stones.
Brand:  Fisherbrand™ FB67851
 View more versions of this product
Product Code. 15505139
  • £1520.00 / 1 unit
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Beschreibung
Beschreibung
  • Class F2 stainless steel calibration weight
  • Weights intended for use in the verification or calibration of class M1 and possibly class M2 weights
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Spezifikation
Spezifikation
F2
1 g to 5 kg
1 g to 5 kg
Cylindrical
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Produktzertifizierungen
Sonderangebote
Sonderangebote