missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Stainless Steel Cylindrical Seamless Scoops

Get optimum results weighing and handling samples and other laboratory materials with cylindrical scoops. Perfect for weighing anything from liquid to gems and other materials, Fisherbrand™ Stainless Steel Seamless Cylindrical Scoops are particularly suitable for pharmaceutical applications.

Brand:  Fisherbrand™ 9035

 View more versions of this product

Product Code. 12981521

  • £147.00 / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Stainless steel seamless cylindrical scoops 
  • Particularly suitable for pharmaceutical applications  
  • All surfaces are non-reacting 
  • Offer zero absorption suiting GMP requirements
  • Scratches within the scoop prevent cross contamination
Specifications

Specifications

Scoop
Autoclavable
Cylindrical
285 mm
Stainless Steel
500 mL
Suitable for Pharmaceutical Applications
120 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.