missing translation for 'onlineSavingsMsg'
Learn More

S Murray™ Stainless Steel Dissection Needle

Use this needle for a variety of applications. S Murray™ Stainless Steel Dissection Needles are fully guaranteed in normal use and subject to stringent quality inspection procedures.

Brand:  S Murray™ D207/01

Product Code. 12760926

  • £38.10 / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Use this needle for a variety of applications. S Murray™ Stainless Steel Dissection Needles are fully guaranteed in normal use and subject to stringent quality inspection procedures.

  • Stainless Steel
  • Length: 130mm
  • Single edged blade (30mm)
Specifications

Specifications

Dissect Needle
Stainless Steel
Straight
130 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.