missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contec™ Stainless Steel Downpress Wringer
Stainless Steel Downpress Wringer for string mops (sold separately).
Brand: Contec™ 2995
752.00 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Contec has a variety of wringers available for curved, flat, and string mops
- Stainless Steel Downpress Wringer 2995 is for use with string mops
- All wringers are fully autoclavable and are compatible with a wide variety of disinfectants
Specifications
Mop Wringer | |
Autoclavable | |
16 in. | |
10 in. | |
Non-sterile | |
25.4 cm |
Stainless Steel | |
25.4 x 25.4 x 40.6 cm (10 x 10 x 16 in.) | |
40.6 cm | |
25.4 cm | |
10 in. |