missing translation for 'onlineSavingsMsg'
Learn More

S Murray™ Stainless Steel Entomological Spring Forceps

Use these forceps for a variety of applications. S Murray™ Stainless Steel Entomological Spring Forceps are fully guaranteed in normal use and subject to stringent quality inspection procedures.

Brand:  S Murray™ E009/01

Code : 03

Additional Details : Weight : 0.01440kg

Product Code. 12770916

  • £11.40 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • Stainless steel
  • With guide pin
  • Length: 120mm
Specifications

Specifications

Entomological Spring Forceps
For dissection
120 mm
Stainless Steel
Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers