Learn More
BD Microlance™ Stainless Steel Needles
Color coded for easy recognition of the needle’s guage. BD™ Microlance™ Stainless Steel Needles feature thin walls for optimal flow and fit Luer slip syringes. These sterile, individually wrapped needles ensure more comfortable insertion with less risk for punching.
Brand:  BD 304727
 View more versions of this product
Product Code. 10595314
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Unique slip angle and polishing method results in less patient discomfort at the injection site and less risk for punching.
  • Hypodermic needles with thin walls for optimal flow
  • Fits syringes connected to a Luer adapter or Luer Lock connector
  • Color coded for quick recognition of gauge for easy selection of appropriate flow rate
  • Sterile and individually wrapped for hygienic, single-use application
  • Stainless steel needles with epoxy resin adhesives
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel with Epoxy Resin Adhesives
5,000 individually wrapped needles, normal cannula wall
0.7 mm
Yes
Needle
SZR -150, connected to a Luer adapter or Luer Lock connector
Black
22 G
For single use only
1.0 in.
Bevel
Sterile