Learn More
BINDER™ Expansion Rack, Includes Calibration
Product Code. 16626483 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers VD 115, VDL 115
BINDER Chambers VD 23, VDL 23
BINDER Chambers VD 56, VDL 56
Unit Size:
Each
Product Code. 16626483
Brand: BINDER™ 86120012
Only null left
Add to basket
Add to basket
Use this stainless steel expansion rack instead of the standard aluminum expansion rack. Includes calibration.
  • Stainless steel
  • Accessory rack
Specifications
Description Expansion racks - stainless steel expansion rack instead of standard aluminum expansion rack, incl. calibration
For Use With (Equipment) BINDER Chambers VD 56, VDL 56
Type Rack
Includes Calibration
Material Stainless Steel