Learn More
Grant Instruments™ Stainless Steel Flask Clamp
Holds flasks in place when used with Grant Aqua Pro Shaking Baths
Brand:  Grant Instruments™ SC-250
 View more versions of this product
Product Code. 15385852
  • £64.20 / Each
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Spring clamp provides security while allowing easy flask insertion/removal
  • For use with OLS and LSB Model Aqua Pro Shaking Water Baths
Specifications
Specifications
250mL Flask
Flask Clamp
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Grant Instruments™ Stainless Steel Flask Clamp > For Use With: 250mL Flask