Learn More
Stainless Steel Foil, 0.25mm (0.01 in.) thick, Thermo Scientific Chemicals
£28.70 - £51.00
Specifications
Chemical Name or Material | Stainless Steel |
---|---|
Color | Silver |
Appearance | Silver -colored foil |
Molecular Formula | Fe:Cr:Ni; 70:19:11 wt% |
MDL Number | MFCD00148499 |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11490401
|
Thermo Scientific Alfa Aesar
041584.FI |
50 x 50 mm | N/A | N/A | N/A | |||||
11328708
|
Thermo Scientific Alfa Aesar
041584.GH |
100 x 100 mm |
£51.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel | |
Silver -colored foil | |
MFCD00148499 | |
Odorless |
Silver | |
Fe:Cr:Ni; 70:19:11 wt% | |
It is insoluble in water. | |
Foil |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.