missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Stainless Steel Freezer Rack
GREENER_CHOICE

Product Code. p-8009467 Shop All Fisherbrand Products
Click to view available options
Shelves:
Pivoting Pull-out Drawers
Vertical Locking Rods
Unit Size:
Each

Product Code. 11786013

Only null left
Add to basket
Add to basket

Store assay plates and PCR racks securely in steel freezer racks. Stainless Steel Freezer Racks are available in two styles: locking rod racks that allow stacking of assay plates or horizontal drawer racks for easy box access. Both styles provide 16 compartments of secure cryostorage.

For cryostorage in upright freezers.

  • Constructed of corrosion-resistant stainless steel.
  • For Assay Plate, PCR rack and cryogenic tube box storage.
  • Designed to fit in most standard upright freezers.
  • Sturdy containers with 4 levels of storage, providing 16 total storage compartments.
  • Wire handles on the front allow for easy removal of the rack from the freezer.

Locking Rod Freezer Rack:

  • Rack includes four vertical locking rods over the side opening to secure contents in place.
  • Allows stacking of short assay plates in each slot (not included):
    • 96-Well Assay plates (holds 3 per compartment, up to 48 total)
    • 24-Well Assay plates (holds 2 per compartment, up to 32 total)
    • 96-Deep Well Assay plates (holds 1 per compartment, up to 16 total)
    • 96-Well PCR racks (holds 1 per compartment, up to 16 total)

Horizontal Drawer Freezer Rack

  • Horizontal drawer system allow for easy box access
  • Drawers feature a stop that pivots down when fully extended
  • Polycarbonate drawer handles provide a user-friendly grip
  • Includes a slot and label for drawer/sample identification
  • Ideal for standard boxes up to 133 x 133 x 53 mm in size

Specifications

Product Type Freezer Rack
Array 4 x 4
Autoclavable Not Autoclavable
Color Silver
Holds Boxes (2 in.)
Material Stainless Steel
Positions 16
Shape Rectangular
Shelves Pivoting Pull-out Drawers
Stackable Yes
Sterility Non-sterile
For Use With (Application) For standard PCR rack and cryogenic rack (up to 133 x 133 x 53 mm) storage in upright freezers
No. of Racks 4
No. per Pack 1 Each
Quantity 1 Ea.
Dimensions (L x W x H) 571 x 139 x 238 mm
Length (Metric) 571 mm
Length (English) 22.5 in.
Width (Metric) 139 mm
Width (English) 5.5 in.
Height (Metric) 238 mm
Height (English) 9.4 in.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.